Merck
  • A chemostat array enables the spatio-temporal analysis of the yeast proteome.
A chemostat array enables the spatio-temporal analysis of the yeast proteome.
Proceedings of the National Academy of Sciences of the United States of America (2013-09-11)
Nicolas Dénervaud, Johannes Becker, Ricard Delgado-Gonzalo, Pascal Damay, Arun S Rajkumar, Michael Unser, David Shore, Felix Naef, Sebastian J Maerkl
ABSTRACT
Observing cellular responses to perturbations is central to generating and testing hypotheses in biology. We developed a massively parallel microchemostat array capable of growing and observing 1,152 yeast-GFP strains on the single-cell level with 20 min time resolution. We measured protein abundance and localization changes in 4,085 GFP-tagged strains in response to methyl methanesulfonate and analyzed 576 GFP strains in five additional conditions for a total of more than 10,000 unique experiments, providing a systematic view of the yeast proteome in flux. We observed that processing bodies formed rapidly and synchronously in response to UV irradiation, and in conjunction with 506 deletion-GFP strains, identified four gene disruptions leading to abnormal ribonucleotide-diphosphate reductase (Rnr4) localization. Our microchemostat platform enables the large-scale interrogation of proteomes in flux and permits the concurrent observation of protein abundance, localization, cell size, and growth parameters on the single-cell level for thousands of microbial cultures in one experiment.
Product Description
Supelco
Methyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methyl methanesulfonate, 99%