Merck
  • Extractive spectrophotometric determination of some anti-inflammatory agents with methylene violet.
Extractive spectrophotometric determination of some anti-inflammatory agents with methylene violet.
The Analyst (1989-04-01)
C S Sastry, A S Prasad Tipirneni, M V Suryanarayana
ABSTRACT
A fairly sensitive spectrophotometric method for the determination of ibuprofen, ketoprofen, piroxicam, diclofenac sodium, mefenamic acid or enfenamic acid in bulk samples and pharmaceutical preparations is described, based on the formation of a chloroform-soluble, coloured ion-association complex between the drug and Methylene Violet at pH 7.6.
Sigma-Aldrich
Methylene Violet (Bernthsen), certified by the BSC, Dye content ≥65 %