Merck
  • Isolation and characterization of a serine protease from the nematophagous fungus, Lecanicillium psalliotae, displaying nematicidal activity.
Isolation and characterization of a serine protease from the nematophagous fungus, Lecanicillium psalliotae, displaying nematicidal activity.
Biotechnology letters (2005-09-01)
Jinkui Yang, Xiaowei Huang, Baoyu Tian, Miao Wang, Qiuhong Niu, Keqin Zhang
ABSTRACT
Lecanicillium psalliotae produced an extracellular protease (Ver112) which was purified to apparent homogeneity giving a single band on SDS-PAGE with a molecular mass of 32 kDa. The optimum activity of Ver112 was at pH 10 and 70 degrees C (over 5 min). The purified protease degraded a broad range of substrates including casein, gelatin, and nematode cuticle with 81% of a nematode (Panagrellus redivivus) being degraded after treating with Ver112 for 12 h. The protease was highly sensitive to PMSF (1 mM) indicating it to be a serine protease. The N-terminal amino acid residues of Ver112 shared a high degree of similarity with other cuticle-degrading proteases from nematophagous fungi which suggests a role in nematode infection.
Sigma-Aldrich
p-Toluenesulfonyl fluoride, 98%