Merck
  • Infraciliary bands in the rumen ophryoscolecid ciliate Ostracodinium gracile (Dogiel, 1925), observed by light microscopy.
Infraciliary bands in the rumen ophryoscolecid ciliate Ostracodinium gracile (Dogiel, 1925), observed by light microscopy.
The Journal of eukaryotic microbiology (1998-12-29)
A Ito, S Imai
ABSTRACT
Morphological features of the rumen ciliate Ostracodinium gracile (Dogiel, 1925) are described from pyridinated silver carbonate-impregnated specimens. Ostracodinium gracile has a characteristic arrangement of infraciliary bands not present in other ophryoscolecid ciliates. Buccal infraciliature is composed of three polybrachykineties. The adoral polybrachykinety does not completely encircle the circumference of the vestibular opening, but arches ventrally from its right to left side. The dorsoadoral polybrachykinety extends laterally along the dorsal side of the vestibular opening. The vestibular polybrachykinety extends along the dorsal wall of the long tubular vestibulum. Dorsal infraciliature consists of the dorsal polybrachykinety that extends laterally along the dorsal side of the body. During binary fission, four primordia, that is ventral, right, left, and dorsal primordia, form in the stomatogenic field and develop into the adoral, dorsoadoral, vestibular, and dorsal polybrachykineties of the opisthe, respectively.
Sigma-Aldrich
Silver carbonate, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver carbonate, 99%
Sigma-Aldrich
Silver carbonate on Celite®, extent of labeling: ~50 wt. % loading