Merck
  • Distribution and strain-dependent formation of volatile metabolites in the genus Ceratocystis.
Distribution and strain-dependent formation of volatile metabolites in the genus Ceratocystis.
Antonie van Leeuwenhoek (1983-11-01)
E Sprecher, H P Hanssen
ABSTRACT
The accumulation of volatile metabolites in cultures of 34 strains comprising ten species of the genus Ceratocystis (Ascomycetes) has been investigated under defined culture conditions. The identified compounds include short-chain alcohols and esters, lower terpenes, terpenoids, and 2-phenylethyl acetate. Certain species can be recognized by a number of common volatiles traced in all strains, although the quantities of these constituents may vary enormously (up to a factor 1:1000) within one species. The formation of some metabolites is restricted to a few strains.
Sigma-Aldrich
6-Methyl-5-hepten-2-one, 99%
Sigma-Aldrich
6-Methyl-5-hepten-2-one, natural, ≥98%, FCC
Sigma-Aldrich
6-Methyl-5-hepten-2-one, ≥98%, FCC, FG