Merck
  • Localization of thiol and disulfide groups in guinea pig spermatozoa during maturation and capacitation using bimane fluorescent labels.
Localization of thiol and disulfide groups in guinea pig spermatozoa during maturation and capacitation using bimane fluorescent labels.
Biology of reproduction (1984-11-01)
T T Huang, N S Kosower, R Yanagimachi
ABSTRACT
The distribution of thiols and disulfides in the guinea pig spermatozoon during maturation and capacitation was studied using both membrane-permeable (mBBr) and impermeable (qBBr) forms of bromobimane, a specific fluorescent probe for thiol groups. In conjunction with the disulfide (SS)-reducing agent dithiothreitol (DTT) and the thiol-alkylating agent N-ethylmaleimide (NEM), quantitative spectrofluorometric measurements of the relative amounts of total thiol (SH) versus SS were performed on cauda epididymal spermatozoa. Under conditions labeling 70% of the reactive thiols, the ratio total SS/SH was 2.4/1.0. Contamination by other cell types prevented similar measurements on spermatozoa at earlier stages of epididymal maturation; thus, the qualitative localization of SH and SS groups in these and in capacitated spermatozoa was visualized using fluorescence microscopy. As spermatozoa moved from the testis to the caput epididymidis, there was a slight apparent increase in staining both on the surface and internally in all regions. Thereafter, surface and internal staining decreased by the time spermatozoa reached the cauda epididymidis. Fluorescence patterns were unaltered under short-term (1 h) capacitation conditions in calcium-free modified Tyrode's medium containing lysophosphatidyl choline and after induction of the acrosome reaction with 2 mM calcium. However, long-term capacitation (16-18 h) in calcium-free modified Tyrode's medium resulted in a loss of detectable SH in the head and acrosome. Regardless of the stage examined, sperm tails contained the greatest relative amount of SH, followed by the head and the acrosome. In addition, there was always more SH detectable internally than on the surface. DTT pretreatment caused a dramatic increase in staining in all regions, both surface and internal, consistent with the quantitative estimates of the SS/SH ratio.
Sigma-Aldrich
Monobromo(trimethylammonio)bimane bromide, suitable for fluorescence, ≥90.0% (HPLC)