Merck
  • Hormonal contraception and HIV-1 infection: medroxyprogesterone acetate suppresses innate and adaptive immune mechanisms.
Hormonal contraception and HIV-1 infection: medroxyprogesterone acetate suppresses innate and adaptive immune mechanisms.
Endocrinology (2013-01-29)
Richard P H Huijbregts, E Scott Helton, Katherine G Michel, Steffanie Sabbaj, Holly E Richter, Paul A Goepfert, Zdenek Hel
ABSTRACT
Recent observational studies indicate an association between the use of hormonal contraceptives and acquisition and transmission of HIV-1. The biological and immunological mechanisms underlying the observed association are unknown. Depot medroxyprogesterone acetate (DMPA) is a progestin-only injectable contraceptive that is commonly used in regions with high HIV-1 prevalence. Here we show that medroxyprogesterone acetate (MPA) suppresses the production of key regulators of cellular and humoral immunity involved in orchestrating the immune response to invading pathogens. MPA inhibited the production of interferon (IFN)-γ, IL-2, IL-4, IL-6, IL-12, TNFα, macrophage inflammatory protein-1α (MIP-1α), and other cytokines and chemokines by peripheral blood cells and activated T cells and reduced the production of IFNα and TNFα by plasmacytoid dendritic cells in response to Toll-like receptor-7, -8, and -9 ligands. Women using DMPA displayed lower levels of IFNα in plasma and genital secretions compared with controls with no hormonal contraception. In addition, MPA prevented the down-regulation of HIV-1 coreceptors CXCR4 and CCR5 on the surface of T cells after activation and increased HIV-1 replication in activated peripheral blood mononuclear cell cultures. The presented results suggest that MPA suppresses both innate and adaptive arms of the immune system resulting in a reduction of host resistance to invading pathogens.
Product Description
Medroxyprogesterone acetate for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Medroxyprogesterone 17-acetate, VETRANAL®, analytical standard
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
Medroxyprogesterone acetate, European Pharmacopoeia (EP) Reference Standard