Merck
  • Conjugated linoleic acid is related to bone mineral density but does not affect parathyroid hormone in men.
Conjugated linoleic acid is related to bone mineral density but does not affect parathyroid hormone in men.
Nutrition research (New York, N.Y.) (2012-12-19)
Jason R Deguire, Nour Makarem, Catherine A Vanstone, Suzanne Morin, Gustavo Duque, Hope A Weiler
ABSTRACT
The relationships between conjugated linoleic acid (CLA) status, bone, body composition, and the effect of CLA on calciotropic hormones are unclear. A cross-sectional study was designed to examine the association between c9, t11 CLA status in erythrocyte membranes (RBC) and body composition. This preceded a dose-response trial investigating if c9, t11 CLA affected parathyroid hormone (PTH). It was hypothesized that (1) higher c9, t11 CLA status in RBC will be associated with a lower fat and higher bone mass and that (2) PTH will be reduced by 30% after supplementation of c9, t11 CLA. Fifty-four men (age, 19-53 years) were included in the cross-sectional analysis, of which 31 were studied in the dose-response trial and randomized to 1 of 3 groups: placebo (n = 10), 1.5 g/d (n = 11), or 3.0 g/d (n = 10) of c9, t11 CLA for 16 weeks. Men with RBC c9, t11 CLA status above the median had higher whole body bone mineral density (BMD) (1.359 ± 0.024 vs 1.287 ± 0.023 g/cm(2); P = .04) and whole body lean mass (WBL) percentage (78.8% ± 0.9% vs 75.3% ± 1.0%; P = .01), whereas body mass index (24.8 ± 0.5 kg/m(2) vs 27.3 ± 0.9 kg/m(2); P = .01) and whole body fat mass percentage (17.3% ± 0.9% vs 21.3% ± 1.1%; P = .007) were lower. In regression analysis, RBC c9, t11 CLA status accounted for a significant proportion (r(2) = 0.10) of the variation in whole body BMD (P = .03). There were no time or treatment differences among any bone or biomarkers of bone metabolism including PTH. These findings indicate that RBC c9, t11 CLA status, a reflection of long-term (~4 months) dietary CLA intake, positively relates to BMD. However, c9, t11 CLA supplementation does not appear to affect PTH in healthy men.
Product Description
Supelco
Conjugated (9Z,11E)-Linoleic acid, analytical standard
Supelco
Conjugated (9E,11E)-Linoleic acid, analytical standard