Merck
  • Two-dimensional infrared spectroscopy reveals the structure of an Evans auxiliary derivative and its SnCl4 Lewis acid complex.
Two-dimensional infrared spectroscopy reveals the structure of an Evans auxiliary derivative and its SnCl4 Lewis acid complex.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-10-04)
Andreas T Messmer, Katharina M Lippert, Sabrina Steinwand, Eliza-Beth W Lerch, Kira Hof, David Ley, Dennis Gerbig, Heike Hausmann, Peter R Schreiner, Jens Bredenbeck
ABSTRACT
Determining the structure of reactive intermediates is the key to understanding reaction mechanisms. To access these structures, a method combining structural sensitivity and high time resolution is required. Here ultrafast polarization-dependent two-dimensional infrared (P2D-IR) spectroscopy is shown to be an excellent complement to commonly used methods such as one-dimensional IR and multidimensional NMR spectroscopy for investigating intermediates. P2D-IR spectroscopy allows structure determination by measuring the angles between vibrational transition dipole moments. The high time resolution makes P2D-IR spectroscopy an attractive method for structure determination in the presence of fast exchange and for short-lived intermediates. The ubiquity of vibrations in molecules ensures broad applicability of the method, particularly in cases in which NMR spectroscopy is challenging due to a low density of active nuclei. Here we illustrate the strengths of P2D-IR by determining the conformation of a Diels-Alder dienophile that carries the Evans auxiliary and its conformational change induced by the complexation with the Lewis acid SnCl(4), which is a catalyst for stereoselective Diels-Alder reactions. We show that P2D-IR in combination with DFT computations can discriminate between the various conformers of the free dienophile N-crotonyloxazolidinone that have been debated before, proving antiperiplanar orientation of the carbonyl groups and s-cis conformation of the crotonyl moiety. P2D-IR unequivocally identifies the coordination and conformation in the catalyst-substrate complex with SnCl(4), even in the presence of exchange that is fast on the NMR time scale. It resolves a chelate with the carbonyl orientation flipped to synperiplanar and s-cis crotonyl configuration as the main species. This work sets the stage for future studies of other catalyst-substrate complexes and intermediates using a combination of P2D-IR spectroscopy and DFT computations.
Sigma-Aldrich
Tin(IV) chloride, 99.995% trace metals basis
Sigma-Aldrich
Tin(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Tin(IV) chloride, 98%