Merck
  • Ginkgolide B reduces atherogenesis and vascular inflammation in ApoE(-/-) mice.
Ginkgolide B reduces atherogenesis and vascular inflammation in ApoE(-/-) mice.
PloS one (2012-06-05)
Xiyun Liu, Gexin Zhao, Yan Yan, Li Bao, Beidong Chen, Ruomei Qi
ABSTRACT
To investigate whether ginkgolide B (a platelet-activating factor inhibitor) affects vascular inflammation in atherosclerosis-prone apolipoprotein E-deficient (ApoE(-/-)) mice. Human platelets were used to evaluate the effects of ginkgolide B on platelet aggregation and signal transduction. Ginkgolide B attenuated platelet aggregation and inhibited phosphatidylinositol 3 kinase (PI3K) activation and Akt phosphorylation in thrombin- and collagen-activated platelets. ApoE(-/-) mice were administered a high-cholesterol diet for 8 weeks. Plasma platelet factor 4 (PF4) and RANTES (regulated upon activation, normal T-cell expressed, and secreted protein) were then measured using an enzyme-linked immunosorbent assay. Scanning electron microscopy and immunohistochemistry were used to determine atherosclerotic lesions. Ginkgolide B decreased plasma PF4 and RANTES levels in ApoE(-/-) mice. Scanning electron microscopic examination showed that ginkgolide B reduced aortic plaque in ApoE(-/-) mice. Immunohistochemistry analysis demonstrated that ginkgolide B diminished P-selectin, PF4, RANTES, and CD40L expression in aortic plaque in ApoE(-/-) mice. Moreover, ginkgolide B suppressed macrophage and vascular cell adhesion protein 1 (VCAM-1) expression in aorta lesions in ApoE(-/-) mice. Similar effects were observed in aspirin-treated ApoE(-/-) mice. Ginkgolide B significantly reduced atherosclerotic lesions and P-selectin, PF4, RANTES, and CD40L expression in aortic plaque in ApoE-/- mice. The efficacy of ginkgolide B was similar to aspirin. These results provide direct evidence that ginkgolide B inhibits atherosclerosis, which may be associated with inhibition of the PI3K/Akt pathway in activated platelets.
Product Description
Supelco
Ginkgolide B from Ginkgo biloba leaves, ≥90% (HPLC)
Supelco
Ginkgolide B, analytical standard