Merck
Bioactive phenolic amides from Celtis africana.
Molecules (Basel, Switzerland) (2012-03-07)
Areej Mohammad Al-Taweel, Shagufta Perveen, Azza Muhammed El-Shafae, Ghada Ahmed Fawzy, Abdul Malik, Nighat Afza, Lubna Iqbal, Mehreen Latif
ABSTRACT
Nine compounds have been isolated for the first time from Celtis africana, namely trans-N-coumaroyltyramine (1), trans-N-feruloyltyramine (2), trans-N-caffeoyltyramine (3), lauric acid (4), oleic acid (5), palmitic acid (6), lupeol (7), β-sitosterol (8) and oleanolic acid (9), respectively. Their structures have been elucidated by different spectroscopic techniques. The isolated compounds were screened for their antioxidant, anti-inflammatory and acetylcholinestrease enzyme inhibitory activities. Compounds 1-3 showed significant antioxidant and anti-inflammatory activities and weak to moderate acetylcholinestrease enzyme inhibition activity.
Product Description
Supelco
Lupeol, analytical standard
Sigma-Aldrich
Lupeol, ≥94%