Merck
  • Branch-selective reductive coupling of 2-vinyl pyridines and imines via rhodium catalyzed C-C bond forming hydrogenation.
Branch-selective reductive coupling of 2-vinyl pyridines and imines via rhodium catalyzed C-C bond forming hydrogenation.
Journal of the American Chemical Society (2008-09-02)
Venukrishnan Komanduri, Christopher D Grant, Michael J Krische
ABSTRACT
Hydrogenation of 2-vinyl azines 1a-1e in the presence of N-arylsulfonyl imines 2a-2l at ambient temperature and pressure employing cationic rhodium catalysts ligated by tri-2-furylphosphine results in regioselective reductive coupling to furnish branched products of imine addition 3a-3v, which embody modest to high levels of syn-diastereoselectivity. Catalytic coupling of 6-bromo-2-vinylpyridine 1a to imine 2l under an atmosphere of elemental deuterium provides deuterio-3l, with deuterium exclusively at the former beta-position of the vinyl moiety. These data are consistent with a catalytic mechanism involving oxidative coupling of the vinyl azine and imine partners to furnish a cationic aza-rhodacyclopentane, which upon deuteriolytic cleavage releases the adduct and regenerates cationic rhodium(I) to close the catalytic cycle. These studies represent the first metal catalyzed reductive C-C couplings of vinyl azines.
Sigma-Aldrich
2-Vinylpyridine, 97%
Sigma-Aldrich
Bis(1,5-cyclooctadiene)rhodium(I) tetrakis[bis(3,5-trifluoromethyl)phenyl]borate