Merck
  • Bilateral chondrodermatitis nodularis chronica helicis on the free border of the helix in a woman.
Bilateral chondrodermatitis nodularis chronica helicis on the free border of the helix in a woman.
Journal of the American Academy of Dermatology (2003-09-27)
Sophia Oelzner, Peter Elsner
ABSTRACT
Chondrodermatitis nodularis chronica helicis is characterized by the presence of small, intensely painful nodules on the free border of the helix of the ear with unknown origin. The lesion is typically localized unilaterally on the right ear. Pressure and trauma may play a role.
Product Description
Clobetasone butyrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clobetasone butyrate, ≥98%
Clobetasone butyrate for system suitability, European Pharmacopoeia (EP) Reference Standard