Merck
  • The entero-insular axis in type 2 diabetes--incretins as therapeutic agents.
The entero-insular axis in type 2 diabetes--incretins as therapeutic agents.
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association (2001-07-20)
W Creutzfeldt
ABSTRACT
The search for intestinal factors regulating the endocrine secretion of the pancreas started soon after the discovery of secretin, i.e. nearly 100 years ago. Insulinotropic factors of the gut released by nutrients and stimulating insulin secretion in physiological concentrations in the presence of elevated blood glucose levels have been named incretins. Of the known gut hormones only gastric inhibitory polypeptide (GIP) and glucagon-like polypeptide-1 (GLP-1 [7-36] amide) fulfill this definition.--The incretin effect (i.e. the ratio between the integrated insulin response to an oral glucose load and an isoglycaemic intravenous glucose infusion) is markedly diminished in patients with type 2 diabetes mellitus, while the plasma levels of GIP and GLP-1 and their responses to nutrients are in the normal range. Therefore, a reduced responsiveness of the islet B-cells to incretins has been postulated. This insensitivity of the diabetic B-cells towards incretins can be overcome by supraphysiological (pharmacological) concentrations of GLP-1 [7-36], however not of GIP. Accordingly, fasting and postprandial glucose levels can be normalized in patients with type 2 diabetes by infusions of GLP-1 [7-36]. Further studies revealed that this is partially due to the fact that GLP-1 [7-36]--in addition to its insulinotropic effect--also inhibits glucagon secretion and delays gastric emptying. These three antidiabetic effects qualify GLP-1 [7-36] as an interesting therapeutic tool, mainly for type 2 diabetes. However, because of its short plasma half life time natural GLP-1 [7-36] is not suitable for subcutaneous application. At present methods are being developed to improve the pharmacokinetics of GLP-1 by inhibition of the cleaving enzyme dipeptidyl peptidase IV (DPP-IV) or by synthesis of DPP-IV resistant GLP-1 analogues. Also naturally occurring GLP-1 analogues (for instance exendin-4) with a much longer half life time than GLP-1 [7-36] are being tested.--Thus, after 100 years of speculations and experimentations, incretins and their analogues are emerging as new antidiabetic drugs.
Sigma-Aldrich
Glucagon-Like Peptide I Amide Fragment 7-36 human, ≥97% (HPLC), powder