Merck
  • Synthesis of 6-deoxy-L-idose and L-acovenose from 1,2: 5,6-di-O-isopropylidene-alpha-D-glucofuranose.
Synthesis of 6-deoxy-L-idose and L-acovenose from 1,2: 5,6-di-O-isopropylidene-alpha-D-glucofuranose.
Carbohydrate research (2001-06-12)
S C Hung, S R Thopate, R Puranik
ABSTRACT
A practical route toward the synthesis of 6-deoxy-L-idose and L-acovenose from 1,2:5,6-di-O-isopropylidene-alpha-D-glucofuranose is described. Key steps include the stereoselective hydrogenation of 6-deoxy-1,2:3,5-di-O-isopropylidene-alpha-D-xylo-hex-5-enofuranose, regioselective protection of 6-deoxy-1,2-O-isopropylidene-beta-L-idofuranose at 0-5, and epimerisation of 6-deoxy-5-O-tert-butyldimethylsilyl-1,2-O-isopropylidene-beta-L-idofuranose at C-3.
Sigma-Aldrich
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose, 98%
Sigma-Aldrich
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose, purum, ≥98.0% (TLC)