Merck
  • Ovarian changes and development of the branchial placenta occurring in Jenynsia lineata (Cyprinodontiformes, Anablepidae).
Ovarian changes and development of the branchial placenta occurring in Jenynsia lineata (Cyprinodontiformes, Anablepidae).
Journal of morphology (2023-08-16)
Luca Di Cesare, Claudio G Barbeito, Carlos Santamaría-Martín, Martín M Montes, Mari C Uribe, Silvia E Plaul
ABSTRACT
In viviparous teleosts, intraovarian gestation occurs intrafollicularly, as in poeciliids, or intraluminally, as in goodeids and anablepids. Furthermore, there are two different forms of embryonic nutrition: lecithotrophy and matrotrophy; depending on the species, these can be exclusive or coexist during gestation. In matrotrophic species, nutrients are transmitted from the mother to the embryo and are especially important in species with intraluminal gestation. Jenynsia lineata is a South American viviparous teleost with intraluminal gestation, characterized by eggs with scarce yolk, which is resorbed when embryos are 6 mm long, thus developing a branchial placenta. Using histological, histochemical, and immunohistochemical techniques, the present study describes the characteristics and changes of the ovarian mucosa in J. lineata during gestational and nongestational phases, and analyzes the embryonic pharyngeal epithelium in the branchial placenta. The ovaries of 30 adult female specimens were processed using histological techniques and stained with hematoxylin-eosin, Masson's trichrome, and Alcian Blue pH 2.5/periodic acid Schiff reagent. To detect cell proliferation, we used antiproliferating cell nuclear antigen antibody. In nonpregnant females, eosinophilic granular cells (EGCs) and lymphocytes were identified in the lamina propria of the tunica mucosa, and melanomacrophage centers (MMCs) and fibroblasts were identified adjacent to tissue debris in the ovarian folds'. In the cellular debris, an embryo in resorption was observed. In pregnant females, the ovarian mucosa has thin vascularization branches entering the opercular chamber of the embryos, in close contact with the forming gill processes, thereby establishing a branchial placenta. Active cell replacement was observed in these ovarian branches. The identification of fibroblasts, lymphocytes, EGCs, and MMCs adjacent to tissue debris could indicate that these cell types are involved in the embryonic resorption process. Considering the new data obtained in this study on the branchial placenta of J. lineata, we conclude that cell proliferation could be involved in the development of maternal-embryonic interaction.
Sigma-Aldrich
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.