Merck
  • Fractalkine Improves the Expression of Endometrium Receptivity-Related Genes and Proteins at Desferrioxamine-Induced Iron Deficiency in HEC-1A Cells.
Fractalkine Improves the Expression of Endometrium Receptivity-Related Genes and Proteins at Desferrioxamine-Induced Iron Deficiency in HEC-1A Cells.
International journal of molecular sciences (2023-05-13)
Edina Pandur, Ramóna Pap, Gergely Jánosa, Adrienn Horváth, Katalin Sipos
ABSTRACT
Fractalkine (CX3CL1/FKN) is a unique chemokine belonging to the CX3C chemokine subclass. FKN exists in two forms: a membrane-bound form expressed by both endometrium cells and trophoblasts thought to be implicated in maternal-fetal interaction and a soluble form expressed by endometrium cells. Endometrium receptivity is crucial in embryo implantation and a complex process regulated by large numbers of proteins, e.g., cytokines, progesterone receptor (PR), SOX-17, prostaglandin receptors (PTGER2), and tissue inhibitors of metalloproteinases (TIMPs). It has also been reported that iron is important in fertility and affects the iron status of the mother. Therefore, iron availability in the embryo contributes to fertilization and pregnancy. In this study, we focused on the effect of iron deficiency on the secreted cytokines (IL-6, IL-1β, leukocyte inhibitory factor, TGF-β), chemokines (IL-8, FKN), and other regulatory proteins (bone morphogenic protein 2, activin, follistatin, PR, SOX-17, prostaglandin E2 receptor, TIMP2), and the modifying effect of FKN on the expression of these proteins, which may improve endometrium receptivity. Endometrial iron deficiency was mediated by desferrioxamine (DFO) treatment of HEC-1A cells. FKN was added to the cells 24 h and 48 h after DFO with or without serum for modelling the possible iron dependence of the alterations. Our findings support the hypothesis that FKN ameliorates the effects of anemia on the receptivity-related genes and proteins in HEC-1A cells by increasing the secretion of the receptivity-related cytokines via the fractalkine receptor (CX3CR1). FKN may contribute to cell proliferation and differentiation by regulating activin, follistatin, and BMP2 expressions, and to implantation by altering the protein levels of PR, SOX-17, PTGER2, and TIMP2. FKN mitigates the negative effect of iron deficiency on the receptivity-related genes and proteins of HEC-1A endometrium cells, suggesting its important role in the regulation of endometrium receptivity.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Anti-SOX17, from rabbit, purified by affinity chromatography