Merck
  • Cyclin-Dependent Kinase 5 Regulates cPLA2 Activity and Neuroinflammation in Parkinson's Disease.
Cyclin-Dependent Kinase 5 Regulates cPLA2 Activity and Neuroinflammation in Parkinson's Disease.
eNeuro (2022-11-10)
Sangita Paul, Saman Fatihi, Srishti Sharma, Rintu Kutum, Raymond Fields, Harish C Pant, Lipi Thukral, Binukumar Bk
ABSTRACT
Hyperactivation of cyclin-dependent kinase 5 (Cdk5) by p25, contributes to neuroinflammation causing neurodegeneration in Parkinson's disease (PD) and Alzheimer's disease. However, the mechanism by which Cdk5 induces neuroinflammation in the PD brain is largely unexplored. Here, we show that Cdk5 phosphorylates cytosolic phospholipase A2 (cPLA2) at Thr-268 and Ser-505 sites lead to its activation and generation of eicosanoid products. Mutational studies using site-directed mutagenesis and molecular simulations show that the architecture of the protein changes on each single-point mutation. Interestingly, double mutations also led to a severe decline in the activity of cPLA2 and to the disruption of its translocation to the plasma membrane. Further, the brain lysates of transgenic PD mouse models show hyperactivation of Cdk5, resulting in enhanced phosphorylation of Thr-268 and Ser-505 of cPLA2 and its heightened activity, confirming the findings observed in the cell culture model of PD. These phosphorylation sites of cPLA2 and Cdk5 could be explored as the future therapeutic targets against neuroinflammation in PD. Further, conjoint transcriptomic analysis of the publicly available human PD datasets strengthens the hypothesis that genes of the arachidonic acid, prostaglandin synthesis, and inflammatory pathways are significantly upregulated in the case of PD patients compared with that of healthy control subjects.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Calnexin, C-Terminal antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-phospho-c-PLA2 (pSer505) antibody produced in rabbit, affinity isolated antibody