Merck
  • Could Moesin Be a New Marker for Indicating Progression in Endometrial Cancer?
Could Moesin Be a New Marker for Indicating Progression in Endometrial Cancer?
Cancer management and research (2022-04-01)
Elif Agacayak, Aysenur Keles, Ugur Deger, Mehmet Sirin Ozcelik, Nurullah Peker, Reyhan Gunduz, Murat Akkus, Huseyin Buyukbayram
ABSTRACT
This study aims to determine an important parameter in progression from pre-invasive lesions of endometrium to endometrial cancer and also evaluate the effect of this parameter on the progression of endometrial cancer. In our study,30 patients with normal endometrial tissue (group 1), 56 patients who had endometrial hyperplasia without atypia (group 2), 36 patients who had endometrial hyperplasia with atypia (group 3), and 63 patients with endometrial cancer (group 4) were included. Age, parity, body-mass index, systemic diseases, and tumor markers of patients were evaluated. Expression levels of Ezrin, Radixin, and Moesin proteins were immunohistochemically evaluated in terms of frequency, intensity, and score value. When we compared hyperplasia cases with or without atypia; frequency, and score value of ezrin expression and frequency, intensity, and score value of moesin expression was significantly higher in patients who had hyperplasia with atypia (p:0.000 p:0.001 p:0.003, p:0.032 p: 0.035 p:0.015 p:0.005, respectively). It was observed that the frequency and score value of moesin expression were significantly higher in patients with endometrial cancer when compared with patients who had hyperplasia with atypia (p:0.003 p:0.045). The frequency of moesin expression was significantly higher in patients who had postoperative mortality (p:0.030 p:0.039). Increased frequency of moesin expression in the preoperative period in patients with atypical hyperplasia should alert the surgeon in terms of malignancy. If the frequency of moesin expression increases in cases with endometrial cancer, the patient should be followed closely in terms of progression in the postoperative period.
Sigma-Aldrich
Monoclonal Anti-Moesin antibody produced in mouse, clone 38/87, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Radixin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Ezrin antibody produced in mouse, clone 3C12, ascites fluid