Merck
  • Multi-walled carbon nanotubes-CoFe2 O4 nanoparticles as a reusable novel peroxymonosulfate activator for degradation of Reactive Black 5.
Multi-walled carbon nanotubes-CoFe2 O4 nanoparticles as a reusable novel peroxymonosulfate activator for degradation of Reactive Black 5.
Water environment research : a research publication of the Water Environment Federation (2020-01-07)
Sara Ashrafi, Nezamaddin Mengelizadeh, Yousef Dadban Shahamat, Mohammad Reza Zare, Mohammad Jalil, Zohreh Berizi, Mohammad Reza Shooshtarian, Ali Parvizimehr, Razieh Zolghadr
ABSTRACT
In this study, CoFe2 O4 nanoparticles supported on multi-walled carbon nanotubes (MWCNTs), as novel peroxymonosulfate (PMS) activator, were synthesized for degradation of Reactive Black 5 (RB5). The results showed that the maximum removal efficiencies of RB5 (100%), chemical oxygen demand (83.12%), and total organic carbon (65.5%) could happen at pH of 7, catalyst dosage of 100 mg/L, PMS dosage of 2 mM, RB5 concentration of 50 mg/L, and time of 30 min. The results of the temperature effect showed that the activation energy (Ea  = 20.92 kJ/mol) for the synthesized catalyst is much lower compared to other studies. The PMS/MWCNTs-CoFe2 O4 system had higher decolorization efficiency and kinetic rates compared to other adsorption and oxidation systems. Quenching experiments proved that RB5 was degraded by sulfate and hydroxyl radicals. The MWCNTs-CoFe2 O4 catalyst showed suitable stability and reusability even after five consecutive catalytic reactions. The continuous treatment of RB5 in real water resources was performed using catalyst packed in a column reactor, and its results showed the high efficiency of the column in the catalytic treatment of the dye at long reaction time. Based on the proposed degradation pathway, the azo bands and the naphthalene structure of RB5 are oxidized to compounds with low molecular weight. PRACTITIONER POINTS: MWCNTs-CoFe2 O4 was used as a novel recyclable catalyst for the activation of peroxymonosulfate and dye degradation. The rate of dye degradation and peroxymonosulfate activation by MWCNTs-CoFe2 O4 was much higher than that of the catalysts alone. Radical SO 4 · - , with contribution percentage of 73.20%, was the main agent for degradation of Reactive Black 5 dye. MWCNTs-CoFe2 O4 in the dye degradation process showed excellent stability and reusability, lower activation energy, and easier separation. The dye degradation products were identified by gas chromatography and UV-vis spectrophotometric analyses, and their degradation pathway was suggested.
Sigma-Aldrich
Wang resin, extent of labeling: 1.0 mmol/g loading