Merck
  • Rapid quantitation of free fatty acids in human plasma by high-performance liquid chromatography.
Rapid quantitation of free fatty acids in human plasma by high-performance liquid chromatography.
Journal of chromatography. B, Biomedical sciences and applications (1998-12-30)
A Mehta, A M Oeser, M G Carlson
ABSTRACT
We report a rapid and sensitive method for separation and quantitation of free fatty acids (FFAs) in human plasma using high-performance liquid chromatography (HPLC). Two established techniques of lipid extraction were investigated and modified to achieve maximal FFA recovery in a reasonably short time period. A modified Dole extraction method exhibited greater recovery (approximately 90%) and short processing times (30 min) compared to the method of Miles et al. Reversed-phase HPLC using UV detection was used for plasma FFA separation and quantitation. Two phenacyl ester derivatives, phenacyl bromide and p-bromophenacyl bromide, were investigated in order to achieve optimal separation of individual plasma FFAs (saturated and unsaturated) with desirable detection limits. Different chromatographic parameters including column temperature, column type and elution profiles (isocratic and gradient) were tested to achieve optimal separation and recovery of fatty acids. Phenacyl bromide esters of plasma fatty acids were best resolved using an octadecylsilyl column with endcapped silanol groups. An isocratic elution method using acetonitrile-water (83:17) at 2 ml/min with UV detection at 242 nm and a column temperature of 45 degrees C was found to optimally resolve the six major free fatty acids present in human plasma (myristic [14:0], palmitic [16:0], palmitoleic [16:1], stearic [18:0], oleic [18:1] and linoleic [18:2]), with a run time of less than 35 min and detection limits in the nmol range. The entire process including plasma extraction, pre-column derivatization, and HPLC quantitation can be completed in approximately 90 min with plasma samples as small as 50 microl. Over a wide physiological range, plasma FFA concentrations determined using our HPLC method agree closely with measurements using established TLC-GC methods (r2 < or = 0.95). In addition, by measuring [14C] or [3H] radioactivity in eluent fractions following HPLC separation of plasma FFA, this method can also quantitate rates of FFA turnover in vivo in human metabolic studies employing isotopic tracers of one or more fatty acids.
Product Description
Supelco
2,4′-Dibromoacetophenone, LiChropur, ≥99.0% (HPLC)