Merck
  • Autophagy Ablation in Adipocytes Induces Insulin Resistance and Reveals Roles for Lipid Peroxide and Nrf2 Signaling in Adipose-Liver Crosstalk.
Autophagy Ablation in Adipocytes Induces Insulin Resistance and Reveals Roles for Lipid Peroxide and Nrf2 Signaling in Adipose-Liver Crosstalk.
Cell reports (2018-11-15)
Jinjin Cai, Karla M Pires, Maroua Ferhat, Bhagirath Chaurasia, Márcio A Buffolo, Rana Smalling, Ashot Sargsyan, Donald L Atkinson, Scott A Summers, Timothy E Graham, Sihem Boudina
ABSTRACT
Autophagy is a homeostatic cellular process involved in the degradation of long-lived or damaged cellular components. The role of autophagy in adipogenesis is well recognized, but its role in mature adipocyte function is largely unknown. We show that the autophagy proteins Atg3 and Atg16L1 are required for proper mitochondrial function in mature adipocytes. In contrast to previous studies, we found that post-developmental ablation of autophagy causes peripheral insulin resistance independently of diet or adiposity. Finally, lack of adipocyte autophagy reveals cross talk between fat and liver, mediated by lipid peroxide-induced Nrf2 signaling. Our data reveal a role for autophagy in preventing lipid peroxide formation and its transfer in insulin-sensitive peripheral tissues.
Sigma-Aldrich
Anti-ATG3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Glycerol Assay Kit, sufficient for 200 colorimetric or fluorometric tests
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution