Merck
Functional Expression of the P2X7 ATP Receptor Requires Eros.
Journal of immunology (Baltimore, Md. : 1950) (2019-12-22)
Yuta Ryoden, Toshihiro Fujii, Katsumori Segawa, Shigekazu Nagata
ABSTRACT
In response to extracellular ATP, the purinergic receptor P2X7 mediates various biological processes, including phosphatidylserine (PtdSer) exposure, phospholipid scrambling, dye uptake, ion transport, and IL-1β production. A genome-wide CRISPR screen for molecules responsible for ATP-induced PtdSer exposure identified a transmembrane protein, essential for reactive oxygen species (Eros), as a necessary component for P2X7 expression. An Eros-null mouse T cell line lost the ability to expose PtdSer, to scramble phospholipids, and to internalize a dye YO-PRO-1 and Ca2+ ions. Eros-null mutation abolished the ability of an LPS-primed human THP-1 macrophage cell line and mouse bone marrow-derived macrophages to secrete IL-1β in response to ATP. Eros is localized to the endoplasmic reticulum and functions as a chaperone for NADPH oxidase components. Similarly, Eros at the endoplasmic reticulum transiently associated with P2X7 to promote the formation of a stable homotrimeric complex of P2X7. These results indicated that Eros acts as a chaperone not only for NADPH oxidase, but also for P2X7, and contributes to the innate immune reaction.
Product Description
Avanti
18:1-06:0 NBD PC, Avanti Research - A Croda Brand
Avanti
18:1-06:0 NBD PC, Avanti Research - A Croda Brand 810132C