Merck
  • Expanding the regioselective enzymatic repertoire: oxidative mono-cleavage of dialkenes catalyzed by Trametes hirsuta.
Expanding the regioselective enzymatic repertoire: oxidative mono-cleavage of dialkenes catalyzed by Trametes hirsuta.
Chemical communications (Cambridge, England) (2012-02-24)
Caroline E Paul, Aashrita Rajagopalan, Iván Lavandera, Vicente Gotor-Fernández, Wolfgang Kroutil, Vicente Gotor
ABSTRACT
The first report of a biocatalytic regioselective oxidative mono-cleavage of dialkenes was successfully achieved employing a cell-free enzyme preparation from Trametes hirsuta at the expense of molecular oxygen. Selected reactions were performed on a preparative scale affording high to excellent conversions and chemoselectivities.
Sigma-Aldrich
4-Formylphenylboronic acid, ≥95.0%