Merck
Pharmacokinetics of cefoperazone in the parturient.
Antimicrobial agents and chemotherapy (1986-12-01)
B Gonik, S Feldman, L K Pickering, C G Doughtie
ABSTRACT
Limited pharmacokinetic data for cefoperazone are available from the parturient. Because cefoperazone has a dual excretory pattern, primarily via the biliary system and secondarily via the kidney, pregnancy-induced physiologic alterations can influence its deposition and clearance. Twelve term parturients receiving cefoperazone prophylaxis after cesarean section were selected for study. After 2 g of cefoperazone was administered for 1 h intravenously, serial blood samples were assayed by high-pressure liquid chromatography. Plasma protein binding of cefoperazone was studied in vitro. The mean peak cefoperazone concentration +/- standard deviation was 169.9 +/- 60.4 micrograms/ml. The mean half-life was 152 min. Total serum clearance was 80.8 +/- 30.8 ml/min. The steady-state volume of distribution was 14.2 +/- 6.0 liters. All subjects had detectable trough levels at the end of the dosage interval, with a mean value of 6.5 +/- 5.2 micrograms/ml. Protein binding of cefoperazone for parturients was 74.3 +/- 10.9%, compared with 87.7 +/- 3.2% in nonpregnant controls (P less than 0.05). These data suggest that cefoperazone deposition can be greatly influenced by pregnancy. However, unlike several other new antimicrobial agents whose excretions are mainly renal, the cefoperazone half-life and thus trough concentration for the parturient more closely resemble that for the nonpregnant subject.
Sigma-Aldrich
Cefoperazone sodium salt, 870 - 1015 μg/mg anhydrous basis