Merck
  • PI(4,5)P2 forms dynamic cortical structures and directs actin distribution as well as polarity in Caenorhabditis elegans embryos.
PI(4,5)P2 forms dynamic cortical structures and directs actin distribution as well as polarity in Caenorhabditis elegans embryos.
Development (Cambridge, England) (2018-05-05)
Melina J Scholze, Kévin S Barbieux, Alessandro De Simone, Mathilde Boumasmoud, Camille C N Süess, Ruijia Wang, Pierre Gönczy
ABSTRACT
Asymmetric division is crucial for embryonic development and stem cell lineages. In the one-cell Caenorhabditis elegans embryo, a contractile cortical actomyosin network contributes to asymmetric division by segregating partitioning-defective (PAR) proteins to discrete cortical domains. In the current study, we found that the plasma membrane lipid phosphatidylinositol 4,5-bisphosphate (PIP2) localizes to polarized dynamic structures in C. elegans zygotes, distributing in a PAR-dependent manner along the anterior-posterior (A-P) embryonic axis. PIP2 cortical structures overlap with F-actin, and coincide with the actin regulators RHO-1 and CDC-42, as well as ECT-2. Particle image velocimetry analysis revealed that PIP2 and F-actin cortical movements are coupled, with PIP2 structures moving slightly ahead of F-actin. Importantly, we established that PIP2 cortical structure formation and movement is actin dependent. Moreover, we found that decreasing or increasing the level of PIP2 resulted in severe F-actin disorganization, revealing interdependence between these components. Furthermore, we determined that PIP2 and F-actin regulate the sizing of PAR cortical domains, including during the maintenance phase of polarization. Overall, our work establishes that a lipid membrane component, PIP2, modulates actin organization and cell polarity in C. elegans embryos.
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
500 g
Estimated to ship on 24 April 2025
$168.00
1 kg
Estimated to ship on 24 April 2025
$311.00
To order products, please contact your local dealer.
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
100 μg
Estimated to ship on 24 April 2025
$822.00
To order products, please contact your local dealer.