Merck
  • CD56-positive lymphocyte infiltration in relation to human papillomavirus association and prognostic significance in oropharyngeal squamous cell carcinoma.
CD56-positive lymphocyte infiltration in relation to human papillomavirus association and prognostic significance in oropharyngeal squamous cell carcinoma.
International journal of cancer (2015-12-15)
Steffen Wagner, Claus Wittekindt, Miriam Reuschenbach, Ben Hennig, Mauran Thevarajah, Nora Würdemann, Elena-Sophie Prigge, Magnus von Knebel Doeberitz, Thomas Dreyer, Stefan Gattenlöhner, Jens Peter Klussmann
ABSTRACT
Human papillomavirus (HPV)-related squamous cell carcinoma of the oropharynx (OSCC) are clinical and biological distinct from their HPV-unrelated counterparts. Patients with HPV-related OSCC display improved prognosis and therefore we investigated possible immune cell infiltrations associated with this tumor phenotype. We retrospectively analyzed a randomly selected cohort of 140 OSCC for presence of immune cells and HPV by immunohistochemistry and PCR followed by bead-based hybridization (Luminex technology). HPV prevalence was 24.3% as determined by positive staining of p16INK4a and detection of high risk HPV-DNA. We found significantly higher numbers of CD56 positive (CD56+) cells in tumor and surrounding microenvironment in HPV-associated compared to HPV-negative OSCC (t-test: p = 0.004 and p = 0.002). For the entire cohort presence of CD56+ cells was associated with increased overall survival independent from HPV (Kaplan-Meier: p = 0.002; Cox regression: p = 0.042). Presence of CD56+ cells also correlated with a better outcome in HPV-negative and especially in HPV-negative OSCC with alcohol consumption ≤ 2 standard drinks per day (Kaplan-Meier: p = 0.05 and p = 0.003). Immunofluorescence localization of granular Granzyme B (GZMB) within CD56+ cells and coexpression of CD16 and CD56 suggests that detected CD56+ cells mainly represent cytotoxic Natural Killer (NK) cells. The fraction of potentially cytotoxic NK cells was significantly higher in HPV-associated compared to HPV-negative OSCC (Mann-Whitney-U-Test: p = 0.011). The elevated abundance and activity of cytotoxic NK cells in OSCC with HPV driven carcinogenesis might contribute to favorable outcome in HPV-related OSCC.
Sigma-Aldrich
Anti-GZMB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution