
Ubiquitin from bovine erythrocytes

Synonym(s):
ATP-dependent proteolytic factor, Ub
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Product Line
Purity
Tissue
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing