
Tris hydrochloride

Synonym(s):
Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride
Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing