Silica Inorganic Sorbent
Synonym(s):
Silica, fumed, Silica, Silicic anhydride, Silicon dioxide, Silicon dioxide amorphous
Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Separation Technique
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing