![S-Adenosyl-L-homocysteine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/199/472/84835ae2-6030-4ec7-83d5-ea21182d9260/640/84835ae2-6030-4ec7-83d5-ea21182d9260.png)
S-Adenosyl-L-homocysteine
![S-Adenosyl-L-homocysteine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/199/472/84835ae2-6030-4ec7-83d5-ea21182d9260/640/84835ae2-6030-4ec7-83d5-ea21182d9260.png)
Synonym(s):
S-(5′-Adenosyl)-L-homocysteine, 5′-Deoxy-S-adenosyl-L-homocysteine, AdoHcy, S-(5′-Deoxyadenosine-5′)-L-homocysteine
Empirical Formula (Hill Notation):
C14H20N6O5S
CAS Number:
Molecular Weight:
384.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing