
Pongamol

Synonym(s):
1-(4-Methoxy-5-benzofuranyl)-3-phenyl-1,3-propanedione
Empirical Formula (Hill Notation):
C18H14O4
CAS Number:
Molecular Weight:
294.30
Products
Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing