
Poly(pyromellitic dianhydride-co-4,4′-oxydianiline), amic acid solution

Synonym(s):
Polyamic acid
Linear Formula:
(C12H12N2O .C10H2O6)x
Products
Brand
Boiling Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-6 of 6
Compare
Product Number
Description
Pricing