
Poly(ethylene glycol) 2-aminoethyl ether acetic acid

Synonym(s):
Amine-PEG-carboxylic acid, Amino-PEG acetic acid, AminoPEG carboxylic acid, Aminopolyoxyethylene acetic acid
Linear Formula:
H2NC2H4O(C2H4O)nCH2CO2H
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing