![Piperacillin Related Compound E pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/605/498/e5b7cf56-fa6d-4a92-9be9-e90c9627144b/640/e5b7cf56-fa6d-4a92-9be9-e90c9627144b.png)
Piperacillin Related Compound E
![Piperacillin Related Compound E pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/605/498/e5b7cf56-fa6d-4a92-9be9-e90c9627144b/640/e5b7cf56-fa6d-4a92-9be9-e90c9627144b.png)
Synonym(s):
N-Ethylpiperazine-2,3-dione
Empirical Formula (Hill Notation):
C6H10N2O2
CAS Number:
Molecular Weight:
142.16
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2