
Oxytocin

Synonym(s):
α-Hypophamine, Oxytocic hormone
Empirical Formula (Hill Notation):
C43H66N12O12S2
CAS Number:
Molecular Weight:
1007.19
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility