
Oxaliplatin Related Compound F

Synonym(s):
[SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-κN,κN′)diiodoplatinum
Empirical Formula (Hill Notation):
C6H14N2I2Pt
CAS Number:
Molecular Weight:
563.08
Products
Brand
Feature
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing