
Octaethylene glycol monododecyl ether

Synonym(s):
C12E8, Dodecyl octaethylene glycol ether, Dodecyloctaglycol, Polyoxyethylene (8) lauryl ether
Linear Formula:
CH3(CH2)11(OCH2CH2)8OH
CAS Number:
Molecular Weight:
538.75
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing