
Naproxen Related Compound K

Synonym(s):
(1RS)-1-(6-Methoxynaphthalen-2-yl)ethanol, α-(6-Methoxy-2-naphthyl)ethanol, (±)-1-(6-Methoxy-2-naphthyl)ethanol, 1-[6-Methoxy-2-naphthyl]hydroxyethane, 2-(1-Hydroxyethyl)-6-methoxynaphthalene, 6-Methoxy-α-methyl-2-naphthalenemethanol
Empirical Formula (Hill Notation):
C13H14O2
CAS Number:
Molecular Weight:
202.25
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing