
myo-Inositol

Synonym(s):
1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol, myo-Inositol, myo-Inositol
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Reference Material Type
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing