Skip to Content
Merck
MOPSO ≥99% (titration)

MOPSO

MOPSO ≥99% (titration)
Synonym(s):
β-Hydroxy-4-morpholinepropanesulfonic acid, 3-Morpholino-2-hydroxypropanesulfonic acid
Empirical Formula (Hill Notation):
C7H15NO5S
CAS Number:
Molecular Weight:
225.26
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing