
Methyl trans-cinnamate

Synonym(s):
trans-Cinnamic acid methyl ester
Linear Formula:
C6H5CH=CHCO2CH3
CAS Number:
Molecular Weight:
162.19
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing