
Methyl gallate

Synonym(s):
Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate
Linear Formula:
(HO)3C6H2CO2CH3
CAS Number:
Molecular Weight:
184.15
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing