
Methoxypolyethylene glycol succinate N-hydroxysuccinimide

Synonym(s):
mono-Methyl polyethylene glycol succinate N-succinimidyl ester, O-Methyl-O′-succinylpolyethylene glycol 5,000 N-succinimidyl ester, PEG-succinimidyl succinate
CAS Number:
Products
Product Category
Brand
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing