![Methionine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/982/493/a22ac878-7a6c-4d28-a85c-98881fb4d7af/640/a22ac878-7a6c-4d28-a85c-98881fb4d7af.png)
Methionine
![Methionine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/982/493/a22ac878-7a6c-4d28-a85c-98881fb4d7af/640/a22ac878-7a6c-4d28-a85c-98881fb4d7af.png)
Synonym(s):
L-Methionine, (S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid
Linear Formula:
CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing