Skip to Content
Merck
Metamitron PESTANAL®, analytical standard

Metamitron

Metamitron PESTANAL®, analytical standard
Synonym(s):
4-Amino-4,5-dihydro-3-methyl-6-phenyl-1,2,4-triazin-5-one
Empirical Formula (Hill Notation):
C10H10N4O
CAS Number:
Molecular Weight:
202.21
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique