
L-tert-Leucinol

Synonym(s):
(S)-2-Amino-3,3-dimethyl-1-butanol
Linear Formula:
(CH3)3CCH(NH2)CH2OH
CAS Number:
Molecular Weight:
117.19
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing