
Indapamide impurity B

Synonym(s):
4-Chloro-N-(2-methyl-1H-indol-1-yl]-3-sulphamoylbenzamide, Dehydroindapamide
Empirical Formula (Hill Notation):
C16H14ClN3O3S
CAS Number:
Molecular Weight:
363.82
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing