
Cidofovir

Synonym(s):
Cidofovir hydrate, (S)-1-[3-hydroxy-2-(phosphonylmethoxy)propyl]cytosine, (S)-HPMPC; Cidovir, HPMPC, Vistide
Empirical Formula (Hill Notation):
C8H14N3O6P · xH2O
CAS Number:
Molecular Weight:
279.19 (anhydrous basis)
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing