
Antimony(III) fluoride

Synonym(s):
Antimony trifluoride, Trifluorostibine, Antimony(III) fluoride
Linear Formula:
SbF3
CAS Number:
Molecular Weight:
178.76
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing